1H,1H,2H,2H-Perfluoroctyltrichlorsilan

Lieferant: CHEMPUR SILANE

Synonyme: Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-trichlorosilane

SILS13125.10 SILS13125.25
SILNSILS13125.10EA 0 EUR
SILNSILS13125.10 SILNSILS13125.25
1H,1H,2H,2H-Perfluoroctyltrichlorsilan
1H,1H,2H,2H-Perfluoroctyltrichlorsilan
Formel: C₈H₄Cl₃F₁₃Si
Molecular Weight: 481.55 g/mol
MDL: MFCD00042363
CAS-Nummer: 78560-45-9
EINECS: 278-947-6
UN: 2987
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR