1H,1H,2H,2H-Perfluoroctyldimethylchlorsilan

Lieferant: CHEMPUR SILANE

Synonyme: Chlorodimethyl(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-n-octyl)silane

SILS13075.5 SILS13075.25
SILNSILS13075.5EA 0 EUR
SILNSILS13075.5 SILNSILS13075.25
1H,1H,2H,2H-Perfluoroctyldimethylchlorsilan
1H,1H,2H,2H-Perfluoroctyldimethylchlorsilan
Formel: C₁₀H₁₀ClF₁₃Si
Molecular Weight: 440.71 g/mol
MDL: MFCD00042359
CAS-Nummer: 102488-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR