Anisomycin (aus Streptomyces griseolus), Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate

M-Clarity™ Qualitätsstufe = MQ300

A9789-100MG A9789-25MG A9789-5MG
SIALA9789-100MGEA 672 EUR
SIALA9789-100MG SIALA9789-25MG SIALA9789-5MG
Anisomycin (aus Streptomyces griseolus), Sigma-Aldrich®
Anisomycin
Formel: C₁₄H₁₉NO₄
Molecular Weight: 265,31 g/mol
MDL: MFCD00077650
CAS-Nummer: 22862-76-6
EINECS: 245-269-7
UN: 3462
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR