1,3-Dimethyl-4-piperidon

Lieferant: FLUOROCHEM

Synonyme: 1,3-Dimethylpiperidin-4-one

011704-25G 011704-5G
FLUO011704-25GEA 97.2 EUR
FLUO011704-25G FLUO011704-5G
1,3-Dimethyl-4-piperidon
1,3-Dimethyl-4-piperidon
Formel: C₇H₁₃NO
Molecular Weight: 127.1800 g/mol
MDL: MFCD00100893
CAS-Nummer: 4629-80-5
EINECS: 225-046-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR