1-Boc-4-(hydroxymethyl)piperidin

Lieferant: CHEMPUR

Synonyme: tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-4-piperidinemethanol, N-BOC-4-Piperidinemethanol

60-0053.1 60-0053.5
CHMP60-0053.1EA 0 EUR
CHMP60-0053.1 CHMP60-0053.5
1-Boc-4-(hydroxymethyl)piperidin
1-Boc-4-(hydroxymethyl)piperidin
Formel: C₁₁H₂₁NO₃
Molecular Weight: 215,29 g/mol
MDL: MFCD02094488
CAS-Nummer: 123855-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR