3,4-(Methylenedioxy)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-benzo-1,3-dioxole, 2-(Benzo[d][1,3]dioxol-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,4-Ethylenedioxybenzene boronic acid, pinacol ester

PN-2440-5G PN-2440-1G
COBBPN-2440-5GEA 236 EUR
COBBPN-2440-5G COBBPN-2440-1G
3,4-(Methylenedioxy)phenylboronsäurepinakolester
3,4-(Methylenedioxy)phenylboronsäurepinakolester
Formel: C₁₃H₁₇BO₄
Molecular Weight: 248,0827 g/mol
MDL: MFCD05663889
CAS-Nummer: 94838-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR