1,4-Benzodioxan-6-boronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,3-Dihydro-1,4-benzodioxine-6-boronic acid
L20296.03
L20296.06
L20296.03EA
56.5
EUR
L20296.03
L20296.06
1,4-Benzodioxan-6-boronsäure 97%
1,4-Benzodioxan-6-boronsäure
Purity: 97%
CAS No.: 164014-95-3
Molecular Formula: C8H9BO4
Formula Weight: 179.97
Beilstein Reference: 7478648
MDL No.: MFCD01009696
Formel:
C₈H₉BO₄ Molecular Weight: 179,97 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01009696 CAS-Nummer: 164014-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...