1,4-Benzodioxan-6-boronsäure
Lieferant: FLUOROCHEM
Synonyme:
2,3-Dihydro-1,4-benzodioxine-6-boronic acid
016099-1G
016099-5G
FLUO016099-1GEA
82.1
EUR
FLUO016099-1G
FLUO016099-5G
1,4-Benzodioxan-6-boronsäure
1,4-Benzodioxan-6-boronsäure
Formel:
C₈H₉BO₄ Molecular Weight: 179,97 g/mol |
MDL:
MFCD01009696 CAS-Nummer: 164014-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...