3,4-(Methylenedioxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3,4-(Methylenedioxy)benzeneboronic acid, Benzo[d][1,3]dioxol-5-ylboronic acid

BB-2440-5G BB-2440-25G BB-2440-1G
COBBBB-2440-5GEA 63.4 EUR
COBBBB-2440-5G COBBBB-2440-25G COBBBB-2440-1G
3,4-(Methylenedioxy)phenylboronsäure
3,4-(Methylenedioxy)phenylboronsäure
Formel: C₇H₇BO₄
Molecular Weight: 165,94 g/mol
MDL: MFCD01009695
CAS-Nummer: 94839-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR