(Cyanmethyl)triphenylphosphoniumchlorid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
A13096.06
A13096.14
A13096.22
A13096.06EA
25.2
EUR
A13096.06
A13096.14
A13096.22
(Cyanmethyl)triphenylphosphoniumchlorid ≥98%
(Cyanomethyl)triphenylphosphonium chloride
Purity: 98+%
CAS No.: 4336-70-3
Molecular Formula: NCCH2P(C6H5)3Cl
Formula Weight: 337.79
Melting Point: ca 270° dec.
Beilstein Reference: 6222047
UN No.: 3439
MDL No.: MFCD00031672
Formel:
C₂₀H₁₇ClNP Molecular Weight: 337,79 g/mol Schmelzpunkt: ∼ 270 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031672 CAS-Nummer: 4336-70-3 EINECS: 224-383-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...