4-Brom-3-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-3-methylpyrazole

HI-1501-25G HI-1501-100G HI-1501-5G
COBBHI-1501-25GEA 71.2 EUR
COBBHI-1501-25G COBBHI-1501-100G COBBHI-1501-5G
4-Brom-3-methyl-1H-pyrazol
4-Bromo-3-methyl-1H-pyrazole
Formel: C₄H₅BrN₂
Molecular Weight: 161,01 g/mol
MDL: MFCD00005241
CAS-Nummer: 13808-64-5
EINECS: 237-456-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR