4-Brom-3-methyl-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Bromo-3-methylpyrazole

M-Clarity™ Qualitätsstufe = MQ200

278238-1G 278238-5G
SIAL278238-1GEA 26 EUR
SIAL278238-1G SIAL278238-5G
4-Brom-3-methyl-1H-pyrazol, Sigma-Aldrich®
4-Bromo-3-methyl-1H-pyrazole
Formel: C₄H₅BrN₂
Molecular Weight: 161,01 g/mol
MDL: MFCD00005241
CAS-Nummer: 13808-64-5
EINECS: 237-456-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR