3-Amino-4-brom-5-methyl-1H-pyrazol Hydrobromid
Lieferant: Thermo Scientific
Synonyme:
4-Bromo-3-methyl-1H-pyrazol-5-amine hydrobromide
KM01090DA
KM01090EA
MAYBKM01090DAEA
0
EUR
MAYBKM01090DA
MAYBKM01090EA
3-Amino-4-brom-5-methyl-1H-pyrazol Hydrobromid
3-Amino-4-brom-5-methyl-1H-pyrazol Hydrobromid
Formel:
C₄H₇Br₂N₃ Molecular Weight: 256.94 g/mol |
MDL:
MFCD00053023 CAS-Nummer: 167683-86-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...