4-Brom-1-(2-(tert-butyldimethylsilyloxy)ethyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1547-5G HI-1547-1G
COBBHI-1547-5GEA 1490 EUR
COBBHI-1547-5G COBBHI-1547-1G
4-Brom-1-(2-(tert-butyldimethylsilyloxy)ethyl)-1H-pyrazol
4-Brom-1-(2-(tert-butyldimethylsilyloxy)ethyl)-1H-pyrazol
Formel: C₁₁H₂₁BrN₂OSi
MDL: MFCD12756450
CAS-Nummer: 1187385-84-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR