1-Ethyl-4-iod-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1598-1G HI-1598-5G
COBBHI-1598-1GEA 0 EUR
COBBHI-1598-1G COBBHI-1598-5G
1-Ethyl-4-iod-1H-pyrazol
1-Ethyl-4-iodo-1H-pyrazole
Formel: C₅H₇IN₂
MDL: MFCD13195706
CAS-Nummer: 172282-34-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR