4-Iod-1-isopropyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Iodo-1-isopropylpyrazole

HC-3260-5G HC-3260-25G HC-3260-1G
COBBHC-3260-5GEA 385 EUR
COBBHC-3260-5G COBBHC-3260-25G COBBHC-3260-1G
4-Iod-1-isopropyl-1H-pyrazol
4-Iod-1-isopropyl-1H-pyrazol
Formel: C₆H₉IN₂
Molecular Weight: 236,0500 g/mol
MDL: MFCD11867882
CAS-Nummer: 313350-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR