4-Iod-1-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1350-5G HI-1350-1G
COBBHI-1350-5GEA 0 EUR
COBBHI-1350-5G COBBHI-1350-1G
4-Iod-1-methyl-1H-pyrazol
4-Iodo-1-methyl-1H-pyrazole
Formel: C₄H₅IN₂
MDL: MFCD05663860
CAS-Nummer: 39806-90-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR