4-Brom-3,5-dimethyl-1H-pyrazol
Lieferant: COMBI-BLOCKS
Synonyme:
4-Bromo-3,5-dimethylpyrazole
HI-1465-5G
HI-1465-25G
HI-1465-100G
COBBHI-1465-5GEA
36.4
EUR
COBBHI-1465-5G
COBBHI-1465-25G
COBBHI-1465-100G
4-Brom-3,5-dimethyl-1H-pyrazol
4-Bromo-3,5-dimethyl-1H-pyrazole
Formel:
C₅H₇BrN₂ Molecular Weight: 175,03 g/mol |
MDL:
MFCD00005242 CAS-Nummer: 3398-16-1 EINECS: 222-263-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...