4-Brom-3,5-dimethyl-1H-pyrazol
Lieferant: FLUOROCHEM
Synonyme:
4-Bromo-3,5-dimethylpyrazole
CFF127.25
CFF127.100
017107-10G
017107-50G
CHMPCFF127.25EA
268
EUR
CHMPCFF127.25
CHMPCFF127.100
FLUO017107-10G
FLUO017107-50G
4-Brom-3,5-dimethyl-1H-pyrazol
4-Bromo-3,5-dimethyl-1H-pyrazole
Formel:
C₅H₇BrN₂ Molecular Weight: 175,03 g/mol |
MDL:
MFCD00005242 CAS-Nummer: 3398-16-1 EINECS: 222-263-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...