4-Brom-1-ethyl-3-methyl-1H-pyrazol-5-carbonylchlorid
Lieferant: FLUOROCHEM
CFF146.1
CFF146.2.5
017217-1G
017217-2.5G
CHMPCFF146.1EA
268
EUR
CHMPCFF146.1
CHMPCFF146.2.5
FLUO017217-1G
FLUO017217-2.5G
4-Brom-1-ethyl-3-methyl-1H-pyrazol-5-carbonylchlorid
4-Brom-1-ethyl-3-methyl-1H-pyrazol-5-carbonylchlorid
Formel:
C₇H₈BrClN₂O |
MDL:
MFCD00173777 CAS-Nummer: 175277-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...