4-Iod-1,3,5-trimethyl-1H-pyrazol

Lieferant: Thermo Scientific

CC14711DA CC14711EA
MAYBCC14711DAEA 0 EUR
MAYBCC14711DA MAYBCC14711EA
4-Iod-1,3,5-trimethyl-1H-pyrazol
4-Iodo-1,3,5-trimethyl-1H-pyrazole
Formel: C₆H₉IN₂
MDL: MFCD00626320
CAS-Nummer: 51660-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR