3-Amino-4-brom-5-phenyl-1H-pyrazol

Lieferant: Thermo Scientific

Synonyme: 4-Bromo-5-phenyl-1H-pyrazol-3-amine, (3-Amino-4-bromo-1H-pyrazol-5-yl)benzene

KM00411DA KM00411EA
MAYBKM00411DAEA 0 EUR
MAYBKM00411DA MAYBKM00411EA
3-Amino-4-brom-5-phenyl-1H-pyrazol
3-Amino-4-bromo-5-phenyl-1H-pyrazole
Formel: C₉H₈BrN₃
Molecular Weight: 238,09 g/mol
MDL: MFCD00082666
CAS-Nummer: 2845-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR