5-Amino-3-(4-bromphenyl)-1H-pyrazol

Lieferant: BIONET RESEARCH

Synonyme: 3-(4-Bromophenyl)-1H-pyrazol-5-amine

5Y-0822.500 5Y-0822.1 5Y-0822.10 5Y-0822.5
BIOT5Y-0822.500EA 0 EUR
BIOT5Y-0822.500 BIOT5Y-0822.1 BIOT5Y-0822.10 BIOT5Y-0822.5
5-Amino-3-(4-bromphenyl)-1H-pyrazol
5-Amino-3-(4-bromphenyl)-1H-pyrazol
Formel: C₉H₈BrN₃
Molecular Weight: 238.09 g/mol
MDL: MFCD00558911
CAS-Nummer: 78583-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR