5-Amino-3-(4-bromphenyl)-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(4-Bromophenyl)-1H-pyrazol-5-amine

M-Clarity™ Qualitätsstufe = MQ100

646733-1G 646733-5G
SIAL646733-1GEA 34.2 EUR
SIAL646733-1G SIAL646733-5G
5-Amino-3-(4-bromphenyl)-1H-pyrazol, Sigma-Aldrich®
5-Amino-3-(4-bromphenyl)-1H-pyrazol
Formel: C₉H₈BrN₃
Molecular Weight: 238,09 g/mol
MDL: MFCD00558911
CAS-Nummer: 78583-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR