5-Pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-(5-Pyrimidyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Pyrimidine-5-boronic acid pinacol ester, Pyrimidyl-5-boronic acid pinacol ester

PN-2360-5G PN-2360-1G
COBBPN-2360-5GEA 158 EUR
COBBPN-2360-5G COBBPN-2360-1G
5-Pyrimidinboronsäurepinakolester
5-Pyrimidinboronsäurepinakolester
Formel: C₁₀H₁₅BN₂O₂
Molecular Weight: 206,05 g/mol
MDL: MFCD05155223
CAS-Nummer: 321724-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR