2-Cyan-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Cyanopyrimidine-5-boronic Acid Pinacol Ester

PN-8748-5G PN-8748-1G
COBBPN-8748-5GEA 961 EUR
COBBPN-8748-5G COBBPN-8748-1G
2-Cyan-5-pyrimidinboronsäurepinakolester
2-Cyan-5-pyrimidinboronsäurepinakolester
Formel: C₁₁H₁₄BN₃O₂
Molecular Weight: 231,0588 g/mol
MDL: MFCD08669562
CAS-Nummer: 1025708-31-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR