2-(Dimethylamino)-5-pyrimidinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
N,N-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
PN-8704-1G
PN-8704-5G
COBBPN-8704-1GEA
127
EUR
COBBPN-8704-1G
COBBPN-8704-5G
2-(Dimethylamino)-5-pyrimidinboronsäurepinakolester
2-(Dimethylamino)-5-pyrimidinboronsäurepinakolester
Formel:
C₁₂H₂₀BN₃O₂ Molecular Weight: 249,12 g/mol |
MDL:
MFCD07368238 CAS-Nummer: 1032759-30-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...