2-(2-Hydroxyethylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-((5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)amino)ethanol

PN-5931-1G PN-5931-250M
COBBPN-5931-1GEA 1140 EUR
COBBPN-5931-1G COBBPN-5931-250M
2-(2-Hydroxyethylamino)-5-pyrimidinboronsäurepinakolester
2-(2-Hydroxyethylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₂H₂₀BN₃O₃
Molecular Weight: 265,1200 g/mol
MDL: MFCD12546562
CAS-Nummer: 1218789-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR