2,4-Dimethoxy-5-pyrimidinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2,4-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2,4-Dimethoxypyrimidine-5-boronic acid pinacol ester
PN-8669-1G
PN-8669-250M
COBBPN-8669-1GEA
497
EUR
COBBPN-8669-1G
COBBPN-8669-250M
2,4-Dimethoxy-5-pyrimidinboronsäurepinakolester
2,4-Dimethoxy-5-pyrimidinboronsäurepinakolester
Formel:
C₁₂H₁₉BN₂O₄ Molecular Weight: 266.1013 g/mol |
MDL:
MFCD06795654 CAS-Nummer: 936250-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...