5-Pyrimidinboronsäure Hemihydrat ≥97%

Lieferant: Thermo Scientific
Achtung

H29212.09 H29212.03
H29212.09EA 276 EUR
H29212.09 H29212.03
5-Pyrimidinboronsäure Hemihydrat ≥97%
5-Pyrimidinboronsäure Hemihydrat

Purity: 97% CAS No.: 696602-91-2 Molecular Formula: C4H5BN2O2.0.5H2O Formula Weight: 132.91 (123.91anhy) Storage Temperature: 2°C to 8°C Melting Point: 300° MDL No.: MFCD03002366

Formel: C₄H₅BN₂O₂·0.5H₂O
Molecular Weight: 132,91 g/mol (123,91 anhy)
Schmelzpunkt: >300 °C
MDL: MFCD03002366
CAS-Nummer: 696602-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR