2-Amino-5-pyrimidinboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Aminopyrimidine-5-boronic acid, (2-Aminopyrimidin-5-yl)boronic acid

M-Clarity™ Qualitätsstufe = MQ100

706221-1G 706221-250MG
SIAL706221-1GEA 193 EUR
SIAL706221-1G SIAL706221-250MG
2-Amino-5-pyrimidinboronsäure, Sigma-Aldrich®
2-Amino-5-pyrimidineboronic acid
Formel: C₄H₆BN₃O₂
Molecular Weight: 138,92 g/mol
MDL: MFCD07375147
CAS-Nummer: 936250-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR