2-Ethoxy-5-pyrimidinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Ethoxypyrimidin-5-yl)boronic acid

BB-9013-5G BB-9013-1G
COBBBB-9013-5GEA 761 EUR
COBBBB-9013-5G COBBBB-9013-1G
2-Ethoxy-5-pyrimidinboronsäure
2-Ethoxy-5-pyrimidineboronic acid
Formel: C₆H₉BN₂O₃
Molecular Weight: 167,9600 g/mol
MDL: MFCD07375134
CAS-Nummer: 1003043-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR