2,4-Dimethoxy-5-pyrimidinboronsäure

Lieferant: COMBI-BLOCKS

BB-8331-5G BB-8331-1G
COBBBB-8331-5GEA 0 EUR
COBBBB-8331-5G COBBBB-8331-1G
2,4-Dimethoxy-5-pyrimidinboronsäure
2,4-Dimethoxy-5-pyrimidineboronic acid
Formel: C₆H₉BN₂O₄
MDL: MFCD01318985
CAS-Nummer: 89641-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR