2-Hydroxy-5-pyrimidinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Oxo-1,2-dihydropyrimidin-5-yl)boronic acid

BB-8780-1G BB-8780-250M
COBBBB-8780-1GEA 385 EUR
COBBBB-8780-1G COBBBB-8780-250M
2-Hydroxy-5-pyrimidinboronsäure
2-Hydroxy-5-pyrimidinboronsäure
Formel: C₄H₅BN₂O₃
Molecular Weight: 139,910 g/mol
MDL: MFCD08275734
CAS-Nummer: 373384-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR