2-Chlor-5-pyrimidinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloropyrimidine-5-boronic acid

BB-5457-5G BB-5457-1G
COBBBB-5457-5GEA 102 EUR
COBBBB-5457-5G COBBBB-5457-1G
2-Chlor-5-pyrimidinboronsäure
2-Chloro-5-pyrimidineboronic acid
Formel: C₄H₄BClN₂O₂
Molecular Weight: 158,35 g/mol
MDL: MFCD08063113
CAS-Nummer: 1003845-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR