4-Isopropyl-5-pyrimidinboronsäure

Lieferant: COMBI-BLOCKS

BB-2600-1G BB-2600-250M
COBBBB-2600-1GEA 1630 EUR
COBBBB-2600-1G COBBBB-2600-250M
4-Isopropyl-5-pyrimidinboronsäure
4-Isopropyl-5-pyrimidinboronsäure
Formel: C₇H₁₁BN₂O₂
Molecular Weight: 165,99 g/mol
MDL: MFCD08235084
CAS-Nummer: 913835-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR