1,3-Dichlor-1,1,3,3-tetraisopropyldisiloxan

Lieferant: CHEMPUR SILANE

Synonyme: 1,3-Dichloro-1,1,3,3-tetrakis(isopropyl)disiloxane

SILS06200.5 SILS06200.25 SILS06200.100 S06200-25G
SILNSILS06200.5EA 0 EUR
SILNSILS06200.5 SILNSILS06200.25 SILNSILS06200.100 FLUOS06200-25G
1,3-Dichlor-1,1,3,3-tetraisopropyldisiloxan
1,3-Dichlor-1,1,3,3-tetraisopropyldisiloxan
Formel: C₁₂H₂₈Cl₂OSi₂
Molecular Weight: 315.43 g/mol
MDL: MFCD00009655
CAS-Nummer: 69304-37-6
UN: 1760
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR