trans,trans-Dibenzylidenaceton
Lieferant: FLUOROCHEM
Synonyme:
(1E,4E)-1,5-Diphenylpenta-1,4-dien-3-one
005548-25G
005548-100G
FLUO005548-25GEA
79.1
EUR
FLUO005548-25G
FLUO005548-100G
trans,trans-Dibenzylidenaceton
trans,trans-Dibenzylidenaceton
Formel:
C₁₇H₁₄O Molecular Weight: 234.30 g/mol |
MDL:
MFCD00004790 CAS-Nummer: 35225-79-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...