trans,trans-Dibenzylidenaceton, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (1E,4E)-1,5-Diphenylpenta-1,4-dien-3-one

M-Clarity™ Qualitätsstufe = MQ200

246425-10G 246425-50G
SIAL246425-10GEA 29.1 EUR
SIAL246425-10G SIAL246425-50G
trans,trans-Dibenzylidenaceton, Sigma-Aldrich®
trans,trans-Dibenzylidenaceton
Formel: C₁₇H₁₄O
Molecular Weight: 234,30 g/mol
MDL: MFCD00004790
CAS-Nummer: 35225-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR