3-(1,3-Oxazol-5-yl)anilin

Lieferant: BIONET RESEARCH

Synonyme: 3-(5-Oxazolyl)aniline

10X-0817.1 10X-0817.10 10X-0817.5 10X-0817.500
BIOT10X-0817.1EA 0 EUR
BIOT10X-0817.1 BIOT10X-0817.10 BIOT10X-0817.5 BIOT10X-0817.500
3-(1,3-Oxazol-5-yl)anilin
3-(1,3-Oxazol-5-yl)anilin
Formel: C₉H₈N₂O
Molecular Weight: 160.1700 g/mol
MDL: MFCD00052191
CAS-Nummer: 157837-31-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR