(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chroman-4-one, 4',5,7-Trihydroxyflavanone, Naringenin

L09834.09 L09834.04
L09834.09EA 114 EUR
L09834.09 L09834.04
(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on ≥97%
(±)-Naringenin

Purity: 97% CAS No.: 67604-48-2 Molecular Formula: C15H12O5 Formula Weight: 272.26 Melting Point: 248-251° Merck Reference: 14,6424 MDL No.: MFCD00006844

Formel: C₁₅H₁₂O₅
Molecular Weight: 272,25 g/mol
Schmelzpunkt: 248…251 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006844
CAS-Nummer: 67604-48-2
EINECS: 266-769-1
Merck Index: 14,6424

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR