N,N-Diethyl-5-borono-2-methoxybenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-(N,N-Diethylsulphamoyl)-4-methoxybenzeneboronic acid

BB-2918-1G BB-2918-5G
COBBBB-2918-1GEA 420 EUR
COBBBB-2918-1G COBBBB-2918-5G
N,N-Diethyl-5-borono-2-methoxybenzolsulfonamid
N,N-Diethyl-5-borono-2-methoxybenzolsulfonamid
Formel: C₁₁H₁₈BNO₅S
MDL: MFCD08056367
CAS-Nummer: 871333-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR