N,N-Dimethyl-2-boronobenzolsulfonamid
Lieferant: COMBI-BLOCKS
Synonyme:
2-(N,N-Dimethylsulphamoyl)benzeneboronic acid
BB-3874-5G
BB-3874-1G
COBBBB-3874-5GEA
811
EUR
COBBBB-3874-5G
COBBBB-3874-1G
N,N-Dimethyl-2-boronobenzolsulfonamid
N,N-Dimethyl-2-boronobenzolsulfonamid
Formel:
C₈H₁₂BNO₄S Molecular Weight: 229,612 g/mol |
MDL:
MFCD04039023 CAS-Nummer: 178432-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...