N,N-Diethyl-3-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-(N,N-Diethylsulphamoyl)benzeneboronic acid

BB-2344-5G BB-2344-1G
COBBBB-2344-5GEA 1180 EUR
COBBBB-2344-5G COBBBB-2344-1G
N,N-Diethyl-3-boronobenzolsulfonamid
N,N-Diethyl-3-boronobenzolsulfonamid
Formel: C₁₀H₁₆BNO₄S
MDL: MFCD07363746
CAS-Nummer: 871329-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR