N,N-Dimethyl-3-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: (3-(N,N-Dimethylsulphamoyl)phenyl)boronic acid, 3-(Dimethylsulphamoyl)benzeneboronic acid

BB-2345-5G BB-2345-1G
COBBBB-2345-5GEA 1690 EUR
COBBBB-2345-5G COBBBB-2345-1G
N,N-Dimethyl-3-boronobenzolsulfonamid
N,N-Dimethyl-3-boronobenzolsulfonamid
Formel: C₈H₁₂BNO₄S
Molecular Weight: 229,06 g/mol
MDL: MFCD07363747
CAS-Nummer: 871329-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR