3-Boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Aminosulphonyl)benzeneboronic acid, 3-Sulphamoylbenzeneboronic acid

BB-2403-1G BB-2403-250M
COBBBB-2403-1GEA 1580 EUR
COBBBB-2403-1G COBBBB-2403-250M
3-Boronobenzolsulfonamid
3-Boronobenzolsulfonamid
Formel: C₆H₈BNO₄S
Molecular Weight: 201.00 g/mol
MDL: MFCD06659840
CAS-Nummer: 850568-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR