N-Butyl-3-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: (3-(N-Butylsulphamoyl)phenyl)boronic acid

BB-2618-5G BB-2618-1G
COBBBB-2618-5GEA 460 EUR
COBBBB-2618-5G COBBBB-2618-1G
N-Butyl-3-boronobenzolsulfonamid
N-Butyl-3-boronobenzolsulfonamid
Formel: C₁₀H₁₆BNO₄S
Molecular Weight: 257.110 g/mol
MDL: MFCD07783858
CAS-Nummer: 871329-78-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR