N-Methyl-3-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Methylsulphamoyl)phenylboronic acid, 3-Methylsulphamoylbenzeneboronic acid

BB-2614-5G BB-2614-1G
COBBBB-2614-5GEA 0 EUR
COBBBB-2614-5G COBBBB-2614-1G
N-Methyl-3-boronobenzolsulfonamid
N-Methyl-3-boronobenzolsulfonamid
Formel: C₇H₁₀BNO₄S
MDL: MFCD07363845
CAS-Nummer: 871329-75-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR