N-tert-Butyl-4-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-(tert-Butylsulphamoyl)benzeneboronic acid, (4-(N-(tert-Butyl)sulphamoyl)phenyl)boronic acid

BB-2406-5G BB-2406-1G
COBBBB-2406-5GEA 612 EUR
COBBBB-2406-5G COBBBB-2406-1G
N-tert-Butyl-4-boronobenzolsulfonamid
N-tert-Butyl-4-boronobenzolsulfonamid
Formel: C₁₀H₁₆BNO₄S
Molecular Weight: 257.11 g/mol
MDL: MFCD06659841
CAS-Nummer: 208516-15-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR