N-Isopropyl-4-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: (4-(N-Isopropylsulphamoyl)phenyl)boronic acid, 4-(Isopropylsulphamoyl)benzeneboronic acid

BB-2427-5G BB-2427-1G
COBBBB-2427-5GEA 886 EUR
COBBBB-2427-5G COBBBB-2427-1G
N-Isopropyl-4-boronobenzolsulfonamid
N-Isopropyl-4-boronobenzolsulfonamid
Formel: C₉H₁₄BNO₄S
Molecular Weight: 243,08 g/mol
MDL: MFCD07363752
CAS-Nummer: 850589-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR